Utility trays, SP Bel-Art
Lieferant: Bel-Art Products, a Part of SP
F163001520 F163001216 F163002020 F163001015 F163002226 F163001818
216-2742EA 435 EUR
216-2742 BELAF163001216 BELAF163002020 BELAF163001015 BELAF163002226 BELAF163001818
Utility trays, SP Bel-Art
Schalen/Schüsseln/Töpfe Laborschalen und Laborwannen
PP, white.
  • Straight sided
  • Temperature resistant up to 135 ºC
  • Easy to clean