Bottle brush, Cleanware™, SP Bel-Art
Lieferant: Bel-Art Products, a Part of SP
F170680000
BELAF170680000EA 150 EUR
BELAF170680000
Bottle brush, Cleanware™, SP Bel-Art
Bürsten
Uniquely designed, swiveling pivot head enables thorough cleaning of bottles and flasks including difficult curves and corners.
  • Natural bristles set in wood will not scratch glass
  • Swivel head fits through a 29/42 standard taper neck, or any non-tapered neck with a diameter of 24 mm (1") or larger
  • Brush head is slightly convex, 45 mm (L) × 21 mm (W) with bristles 8 mm long
  • Brush head is attached to hardwood handle 37 cm long × 7 mm diameter