Pipette jar
Lieferant: Bel-Art Products, a Part of SP
F171300018
BELAF171300018EA 415 EUR
BELAF171300018
Pipette jar
Dosen Pipettenbehälter
Store and soak pipettes in these leakproof, molded polyethylene pipette jars.
Withstands all common cleaning solutions at room temperature, including sulfuric acid/dichromate.