Test tube racks, In-Line, SP Bel-Art
Lieferant: Bel-Art Products, a Part of SP
F185270000
BELAF185270000EA 37.6 EUR
BELAF185270000
Test tube racks, In-Line, SP Bel-Art
Gestelle Reagenzglasgestelle
For high-visibility, six-place high-density polyethylene rack is end-supported by two columns.
  • Top plate has 25 mm diameter holes
  • Base has corresponding hemispherical wells
  • Includes six vertical pins for tube drying
  • Easy snap together assembly