Racks for test tubes, Poxygrid®, SP Bel-Art
Lieferant: Bel-Art Products, a Part of SP
F187650001
BELAF187650001EA 137 EUR
BELAF187650001
Racks for test tubes, Poxygrid®, SP Bel-Art
Gestelle Reagenzglasgestelle
Steel wire, epoxy coating.
  • Chemical and corrosion-resistant
  • Temperature resists from –210 to 121 °C
  • The open-mesh wire design allows fast sample temperature changes when cycling through different water bath temperatures
Steel wire construction with heat-cured epoxy coating is unaffected by organic solvents, salts, weak acids, and weak alkalis. They have square holes, three decks, and a slotted grid bottom and reinforced edges.