Kryoboxen, Scienceware®
Lieferant: Bel-Art Products, a Part of SP
F188490001
BELAF188490001EA 128 EUR
BELAF188490001
Kryoboxen, Scienceware®
Boxen Kryoboxen
These durable polycarbonate boxes are ideal for compact storage of 1,2 - 2,0 or 5 ml cryogenic vials.
  • Temperatures resistance from –196 to +121 °C
  • Clear lids can be written on with permanent markers and have a printed grid for easy inventory
  • Keyed corner aligns the lid with identifying numbers on the rack
  • Can be used as mailers for vials