Tube rack, for water bath, SP Bel-Art
Lieferant: Bel-Art Products, a Part of SP
F188820000
BELAF188820000EA 123 EUR
BELAF188820000
Tube rack, for water bath, SP Bel-Art
Gestelle Reagenzglasgestelle
Height-adjustable racks securely hold up to six tubes or vials at any desired depth in a water bath or beaker. The rack has a flexible grip that easily retains mixed test tubes of varying weight and volume from 1,5 ml to 15 ml.
  • Made of PP
  • Adjustable height prevents submersion problems
  • Compact footprint
  • Stable base
The compact yet stable footprint saves bench and bath space and lifts in and out of a bath without splashing.