Test tube rack slanter, SP Bel-Art
Lieferant: Bel-Art Products, a Part of SP
F188900100
BELAF188900100EA 80.8 EUR
BELAF188900100
Test tube rack slanter, SP Bel-Art
Gestelle Reagenzglasgestelle
Made of PP.
  • Base legs have predetermined slots that stand slides in and out of to easily change angles
  • Fits racks up to 293×242×83 mm
  • Autoclavable
These rack slanters used to tilt the test tube rack to 25°, 35° or 45° to reduce hand strain when working with pipettors. It is also ideal for creating agar slant tubes.