PCR tube rack with cover, SP Bel-Art
Lieferant: Bel-Art Products, a Part of SP
F189020005
BELAF189020005EA 239 EUR
BELAF189020005
PCR tube rack with cover, SP Bel-Art
Gestelle Reagenzglasgestelle
PP, with translucent PP cover.
Designed for preparing and storing 0,2 ml thin-walled PCR tubes and strip tubes before or after PCR sample processing applications. The compact rack features 96 wells that keep tubes securely upright in an 8×12 array. An alphanumeric grid assists in easy sample identification.
W×D×H: 127×89×34 mm
Bestellinformation: The Assortment pack includes one of each colour; flourescent blue, green, pink, yellow and orange.