Pipettor holder system, PiRack™, SP Bel-Art
Lieferant: Bel-Art Products, a Part of SP
F189540000
BELAF189540000EA 94.3 EUR
BELAF189540000
Pipettor holder system, PiRack™, SP Bel-Art
Pipettenständer
This PP system is strong enough to support the weight of even the heaviest pipettor. Hold a single pipette with one clamp or add a triple clamp or extenders to increase the number of holders at a work location.
  • Utility tray mounts between two clamps; ideal for storing pipetting supplies or other small items
  • Attach to any shelf or benchtop edge up to 2,54 cm (1") thick using the large locking thumbscrew clamp
  • Supports many standard brands of pipettors
  • All components are chemical resistant and autoclavable at 121 °C
Bestellen
TECHNISCHE DATEN
Environmentally Preferable