Tray, for Petri dish incubation, SP Bel-Art
Lieferant: Bel-Art Products, a Part of SP
F189830000
BELAF189830000EA 211 EUR
BELAF189830000
Tray, for Petri dish incubation, SP Bel-Art
Schalen/Schüsseln/Töpfe Laborschalen und Laborwannen
Tray made of white epoxy-coated steel.
  • Excellent aid for keeping cell cultures organised
  • With 29 holes of 19 mm diameter in bottom allow air passage
  • Holds 100 mm petri dishes securely on an incubator rack