Contact plate racks, SP Bel-Art
Lieferant: Bel-Art Products, a Part of SP
F189900000
BELAF189900000EA 200 EUR
BELAF189900000
Contact plate racks, SP Bel-Art
Gestelle Spender für Petrischalen
PP moulded rack, ideal for high volume sampling processes.
  • Transport and incubate contact plates and organise bench top sample processing
  • Racks hold 60 contact plates with covers, configured in six columns of ten plates
  • Interlocking when stacked
  • Large centre divider serves as a handle that does not interfere with samples and provides space for labelling
  • Autoclavable at 121 °C
Some simple assembly required. Dimensions provide stability and conserve bench space: 266×165×178 mm.
Four fully loaded racks can be safely stacked to maximise incubator space.
Access slots underneath each stack permits secure removal of a complete stack of plates from the rack using a fixed grip that places a finger under the bottom plate and the thumb on top of the cover of the top plate.