Tubing connectors, Y-shaped, SP Bel-Art
Lieferant: Bel-Art Products, a Part of SP
F196140000 F196130000 F196150000 F196160000 F196180000
BELAF196140000EA 142 EUR
BELAF196140000 BELAF196130000 BELAF196150000 BELAF196160000 BELAF196180000
Tubing connectors, Y-shaped, SP Bel-Art
Fittings und Verbinder
Y-shaped connectors for tubing, made of chemical-resistant polypropylene, for use with a wide range of liquids or gases.
  • Multiple barbs per arm provide additional contact between the fitting and the tubing for extra gripping power
  • FDA-grade for direct contact with food, drugs, and cosmetics
  • Autoclavable