Vacuum aspiration systems, SP Bel-Art
Lieferant: Bel-Art Products, a Part of SP
F199170250
BELAF199170250EA 3320 EUR
BELAF199170250
Vacuum aspiration systems, SP Bel-Art
Pumpen Vakuum-Absaugsysteme
These system is used to aspirate tissue culture media, supernatants, and chemical wastes.
  • Light weight, compact and easy to use; practical whenever a house vacuum is absent or not working
  • Hydrophobic filter protects pump from overflow
  • Low noise and minimal vibration
  • Chemical and corrosion resistant polypropylene collection bottle features heavy wall construction and a safety release vent to prevent vacuum implosion
  • Collection bottle, lid, tubing and connectors are all autoclavable at 121 °C; all held within a polypropylene pump housing
  • Hi-flow model has a stronger flow rate to work reliably with up to 8-channels, even with 4 channels empty
  • Hi-flow model includes quick disconnects to allow easy removal, emptying and replacement of the bottle
Built-in, maintenance free pump quickly and conveniently aspirates fluids.
Standards: Hi-flow model ONLY (BELAF199170250) is CSA certified and the transformer includes an additional cord for 220 V type C outlets.
Lieferumfang: Complete system includes polypropylene bottle (3,78 L), tubing, safety vent and vaccum pump.
Bitte beachten Disposable pipette tip, pasteur pipette needs to be connect to collection tubing and plug into 110/120 V outlet.