Spill containment tray, SP Bel-Art
Lieferant: Bel-Art Products, a Part of SP
F246750050
BELAF246750050EA 199 EUR
BELAF246750050
Spill containment tray, SP Bel-Art
Schalen/Schüsseln/Töpfe Auffangschalen und Auffangbehälter
Made of Polystyrene, translucent.
  • With external flange
  • Durable
  • Liners must be ordered seperately
Protect laboratory benches. To absorb spilled liquids.