Working tray, benchtop, SP Bel-Art
Lieferant: Bel-Art Products, a Part of SP
F246750060
BELAF246750060EA 323 EUR
BELAF246750060
Working tray, benchtop, SP Bel-Art
Schalen/Schüsseln/Töpfe Auffangschalen und Auffangbehälter
Made of Polypropylene.
  • Can be used in combination with biohazard, radioactive or general purpose mats
  • Tray lip assures containment of spilled liquids that are not absorbed by the mats
  • Autoclavable
This plastic tray is designed to hold the most popular sizes of safety mats to help contain spills.