Beta radiation box with lid, SP Bel-Art
Lieferant: Bel-Art Products, a Part of SP
F249830000
BELAF249830000EA 527 EUR
BELAF249830000
Beta radiation box with lid, SP Bel-Art
Boxen Beta-/Gammastrahlen-Sicherheitsbehälter
Acrylic, clear.
  • Excellent visibility of contents
  • Includes removable lid and non-skid feet
This Beta box is made from 9,5 mm (⅜") clear acrylic with tightfitting joints that reduce exposure to radioactive isotopes.