SP Bel-Art tubing sinkers
Lieferant: Bel-Art Products, a Part of SP
F329030100 F329030104 F329030102
BELAF329030100EA 144 EUR
BELAF329030100 BELAF329030104 BELAF329030102
SP Bel-Art tubing sinkers
These Teflon® sinkers help to keep tubing at the bottom of the container.
  • Ideal for use with pumps and dispensers
  • Hose barb ends for simple tubing connection
  • Solvent resistant and autoclavable