Scoops, rectangular, SP Bel-Art
Lieferant: Bel-Art Products, a Part of SP
F367580000
BELAF367580000EA 0 EUR
BELAF367580000
Scoops, rectangular, SP Bel-Art
Schaufeln
PP, rectangular. These sampling scoops are designed for retrieving samples from a larger source. Excellent for scooping anything from bulk powder to ice.
  • Looped handle for hanging
  • Rounded corners for easy cleaning
  • Holds approximately 2 liters (½ gallon)
  • Made from rigid PP