Magnetic stirring bars, PTFE, Spinvane®, SP Bel-Art
Lieferant: Bel-Art Products, a Part of SP
F371350000
BELAF371350000EA 27.4 EUR
BELAF371350000
Magnetic stirring bars, PTFE, Spinvane®, SP Bel-Art
Rührer Magnetrührstäbchen
Spinvane® magnetic stirring bars are made of PTFE and have a cross-mounted, moulded-in Alnico magnet.
  • Triangular shape bar is designed for small conical-shaped tubes and micro vials
  • Half-round tapered shape is designed for conical bottom centrifuge tubes - can be trimmed to fit
  • Half-round shape is designed for Ø16 mm test tubes - can be trimmed to fit
  • Autoclave safe
Standards: Manufactured in a registered ISO 9001:2015 facility, as verified by SGS certification