Magnetic stirring bars, PTFE, for spectrophotometer cells or cuvettes, Spinbar®, SP Bel-Art
Lieferant: Bel-Art Products, a Part of SP
F371500000
BELAF371500000EA 49.7 EUR
BELAF371500000
Magnetic stirring bars, PTFE, for spectrophotometer cells or cuvettes, Spinbar®, SP Bel-Art
Rührer Magnetrührstäbchen
This magnetic stirring bar, with Alnico magnet encapsulated in PTFE coating, fits into the standard 10 mm light path cuvette and provides rapid vertical and horizontal mixing with a minimum of vortexing when placed on a magnetic stirring machine.
  • Centrifugal pumping action, generated by the cross channels in the upper face, mixes without aeration
  • Photometric measurements can proceed uninterrupted while reagents are added
  • One-piece moulded unit will not scratch cuvettes; may also be used in test tubes
  • Autoclavable
Standards: Manufactured in a registered ISO 9001:2015 facility, as verified by SGS certification