Magnetic stirring bars, octagonal, rare earth, PTFE, Spinbar®, SP Bel-Art
Lieferant: Bel-Art Products, a Part of SP
F371740000
BELAF371740000EA
294
EUR
BELAF371740000
Magnetic stirring bars, octagonal, rare earth, PTFE, Spinbar®, SP Bel-Art
Rührer
Magnetrührstäbchen