Magnetic stirring bars, octagonal, rare earth, PTFE, Spinbar®, SP Bel-Art
Lieferant: Bel-Art Products, a Part of SP
F371740000
BELAF371740000EA 294 EUR
BELAF371740000
Magnetic stirring bars, octagonal, rare earth, PTFE, Spinbar®, SP Bel-Art
Rührer Magnetrührstäbchen
PTFE covered samarium cobalt magnet provides strong coupling with drive magnets making it ideal for use with viscous solutions and efficient, high speed stirring. The fluted octagonal shape creates torque transmission rates to the stirred liquid which are two to three fold higher compared to similarly sized standard stirring bars.
  • Eight sided, faceted shape for exceptional mixing power at high speeds
  • Very effective and highly stable in vessels with curved bottoms
  • Demagnetisation due to ambient magnetic fields is eliminated
  • Recessed flutes and distinct edges provide excellent surface area
Standards: Manufactured in a registered ISO 9001:2008 environment, as verified by SGS certification.