Pipette aid, SP Bel-Art
Lieferant: Bel-Art Products, a Part of SP
F378860000
BELAF378860000EA 133 EUR
BELAF378860000
Pipette aid, SP Bel-Art
Pipetten Pipettierbälle
This pipette aid is used by squeezing, then releasing the bulb to aspirate fluid into the pipette.
  • Quickly cover pipette top with a gloved finger to control and dispense fluid
  • Vikem® vinyl bulb has a tapered polyethylene chuck for quick removal when pipette is filled