Pipette controller, economy, Pipette Pump™ III, SP Bel-Art
Lieferant: Bel-Art Products, a Part of SP
F379020002 F379020010 F379020025
BELAF379020002EA 95.7 EUR
BELAF379020002 BELAF379020010 BELAF379020025
Pipette controller, economy, Pipette Pump™ III, SP Bel-Art
Pipetten Pipettierhelfer
These lightweight pipetting devices made of PP are designed for quick and accurate one-handed operation. Available in three colour-coded pipette volumes.
  • Allows arm to remain in a more natural, lowered position, unlike other pipettors that require arm and elbow elevation for many pipetting actions
  • Optimally located thumb wheel enables a relaxed grip and ease of rotation for precision aspirating or dispensing down to single drops; can be used to blow out the pipette
  • Self-stabilising collar inside the silicone chuck holds either glass or plastic pipettes
  • Compact design adds only 15,2 cm to the pipette length, allowing use in fume hoods and controlled environment glove boxes
  • Resistant to acids, alkalis, and most solvents; can be disassembled for cleaning