Pipette controller, Pipette Pump™ III, SP Bel-Art
Lieferant: Bel-Art Products, a Part of SP
F379040025
BELAF379040025EA 97.6 EUR
BELAF379040025
Pipette controller, Pipette Pump™ III, SP Bel-Art
Pipetten Pipettierhelfer
These lightweight pipetting devices made of PP can be used with glass and plastic pipettes up to 2, 10 or 25 ml. Designed for quick and accurate one-handed operation.
  • Fast-release trigger dispenses the pipettes entire contents
  • The ergonomic design enhances comfort, utility, and dispensing precision
  • Allows arm to remain in a more natural, lowered position, unlike other pipettors that require arm and elbow elevation for many pipetting actions
  • Optimally located thumb wheel enables a relaxed grip and ease of rotation for precision aspirating or dispensing down to single drops; can be used to blow out the pipette
  • Self-stabilising collar inside the silicone chuck holds either glass or plastic pipettes
  • Compact design adds only 15,2 cm (6") to the pipette length, allowing use in fume hoods and controlled environment glove boxes
  • Resistant to acids, alkalis, and most solvents; can be disassembled for cleaning