Küvettenspüler Vakuwash®
Lieferant: Bel-Art Products, a Part of SP
F389600000
BELAF389600000EA 423 EUR
BELAF389600000
Küvettenspüler Vakuwash®
Reinigungs- und Desinfektionsautomaten
Economical, durable and gentle all-plastic washer for delicate glass or quartz cuvettes.
  • Mouth of an inverted cuvette is pressed against the rubber cushion to induce suction
  • Water or other cleaning fluid poured into the 89 mm (3½") Ø funnel is forcibly sprayed into the cuvette and then drains into the flask
  • Continued pressure on the cuvette creates an air flow which dries it
  • Final wash in acetone aids in removing water droplets
  • Minimum cuvette Int.Ø must be 9 mm or larger to form a seal over the spray nozzle
As long as the cuvette maintains pressure against the cushion, the continued force sets up a stream of air flow into the cuvette and drys it thoroughly. Any residual water droplets may be removed with a final rinse with acetone.
Lieferumfang: Supplied with 500 ml PP vacuum flask, Vikem® vinyl stopper and 3' PVC tubing ready for connection to a vacuum line.