Evaporating dishes, PP, SP Bel-Art
Lieferant: Bel-Art Products, a Part of SP
F409250000
BELAF409250000EA 108 EUR
BELAF409250000
Evaporating dishes, PP, SP Bel-Art
Schalen/Schüsseln/Töpfe Abdampfschalen
Dishes, made of PP, translucent, smooth, with pour spout.
  • Useful for titration and evaporation
  • Resists acids and alkalis
  • Very good temperature resistance
Can also be used as a reusable weighing dish.