Desiccator cabinets, auto, horizontal, secador®, SP Bel-Art
Lieferant: Bel-Art Products, a Part of SP
F420740220
BELAF420740220EA 3150 EUR
BELAF420740220
Desiccator cabinets, auto, horizontal, secador®, SP Bel-Art
Exsikkatoren Trockenkammern
Secador® 4.0 auto desiccator cabinets are made from moulded Durastar® co-polyester which blocks 99% of UV light and resists staining, crazing and chemical attack.
  • Large, pull down, oven style door with easy-to-view digital hygrometer
  • Two perforated shelves facilitate circulation of dry air
  • Colour-indicating silica gel allows quick visual confirmation of desiccant status
Protecting valuable humidity-sensitive items, such as reagents or electronics, is easy and fully automatic with Secador® Auto-Desiccator cabinets. Automatic dehumidification and desiccant regeneration cycle operates continuously every 20 minutes. Compared to other auto-desiccators that regenerate once every 4 to 6 hours, the Secador® Auto-Desiccators attains a low %rH level on start-up and consistently maintains low %rH, especially with frequent entry to the desiccator interior.
Lieferumfang: Supplied with power cord/transformer with input connector.