Laboratory fume hoods, clear view, SP Bel-Art
Lieferant: Bel-Art Products, a Part of SP
F500201010
BELAF500201010EA 6080 EUR
BELAF500201010
Laboratory fume hoods, clear view, SP Bel-Art
Dunstabzugshauben und Verkleidungen Chemikalien-Abzüge
Scienceware® Clear View fume hoods are suitable for potentially hazardous fumes and dust. It is constructed of polycarbonate with sleek modern style.
  • Perfect for laboratory, classroom or industrial applications
  • Visibility into the fume hood is possible from all sides
  • Three position sliding sash can be completely lowered or set at a choice of two intermediary heights
  • Opening (152 mm, O.D) at the top allows easy connection to exhaust systems