Carboys, space saver, with screw cap, SP Bel-Art
Lieferant: Bel-Art Products, a Part of SP
H109390005
BELAH109390005EA 289 EUR
BELAH109390005
Carboys, space saver, with screw cap, SP Bel-Art
Flaschen Ballons
PP, translucent.
  • In addition to the top handle, a convenient grip hold in the front facilitates removal from a shelf
  • Graduated markings on front in liters
  • Good chemical and temperature resistance
Space saver book bottle stacks book-like on shelves for efficient use of space.