Dispensing jug, SP Bel-Art
Lieferant: Bel-Art Products, a Part of SP
H118500000
BELAH118500000EA 162 EUR
BELAH118500000
Dispensing jug, SP Bel-Art
Flaschen Ballons
LDPE, translucent.
  • Strong and lightweight
  • Storage capacity is 20 L (5 gallons)
  • Gradations in gallons are moulded into the side
Use this jug to store and dispense common solutions, including janitorial cleaners, deionized/distilled water, or other aqueous liquids. Threaded LDPE spigot extends 125 mm from jug and features a 35 mm long stem for dispensing.