Sample bags, SP Bel-Art
Lieferant: Bel-Art Products, a Part of SP
H131780812 H131761016 F131760816 F131761220
BELAH131780812EA 108 EUR
BELAH131780812 BELAH131761016 BELAF131760816 BELAF131761220
Sample bags, SP Bel-Art
Beutel Probenbeutel
These utility reusable, transparent PE bags have variety of applications such as they are ideal for shipping, storage and are especially useful as sample bags.
  • Not autoclavable
  • Latex-free
  • Available in two weights
Standards: FDA approved.