Gas lighter, Piezoelectric, SP Bel-Art
Lieferant: Bel-Art Products, a Part of SP
H150560000
BELAH150560000EA 196 EUR
BELAH150560000
Gas lighter, Piezoelectric, SP Bel-Art
Heizgeräte Gasbrenner
For lighting Bunsen burners, gas stoves, propane or oxy-acetylene torches safely.
  • Does not require batteries or fuel refills
  • Comfortable plastic pistol-grip with push-button trigger creates an electrical spark (not a flame) at 20 kv with each squeeze
Length: 265 mm
Weight: 88 g