Sample containers, SP Bel-Art
Lieferant: Bel-Art Products, a Part of SP
H178760000
BELAH178760000EA 259 EUR
BELAH178760000
Sample containers, SP Bel-Art
Probenbehälter Universalbehälter
LDPE, with screw cap.
  • Smooth walls do not collect particulate and are easy to clean
  • Screw-cap design features a leak proof inner seal
  • Heavy-wall, injection molded strength
Ideal for storing small quantities of liquids or solids. These heavy-walled, injection molded jars have even been used to transport dust samples from the moon. Easy-to-clean with a screw cap design featuring a leak-proof inner seal.