Retort stand, SP Bel-Art
Lieferant: Bel-Art Products, a Part of SP
H183030005
BELAH183030005EA 224 EUR
BELAH183030005
Retort stand, SP Bel-Art
Stative
PP base, stainless steel mounting rod. This non corrosive, molded support stand is an excellent alternative to metal stands.
  • Compact size saves bench space
  • Two rod positions for mounting at center or one end of base
  • Stainless steel mounting rod: Ø 9,4 mm × 50 cm length
  • Hex nut secures rod and prevents it from turning
  • Autoclavable
The compact thick base has a smooth surface for easy cleaning and weighs 1,95 kg for stability.
Bestellinformation: Funnel, ring clamp and flask are not included.