Bosshead, SP Bel-Art
Lieferant: Bel-Art Products, a Part of SP
H183500000
BELAH183500000EA 142 EUR
BELAH183500000
Bosshead, SP Bel-Art
Klemmen Stativmuffen
Stainless steel. This open-sided bosshead accepts rods up to 13 mm diameter.
Holds rods at right angles to one another with flat wing-head clamping screws.