Drum pumps for liquids, SP Bel-Art
Lieferant: Bel-Art Products, a Part of SP
H328870000
BELAH328870000EA 0 EUR
BELAH328870000
Drum pumps for liquids, SP Bel-Art
Pumpen Fass/Transferpumpen
Fits directly to a standard drum bung or carboy. It is constructed entirely of rugged PE and it is resistance to strong acids, alkalis and many solvents. The pump will efficiently deliver against a 3,65 m head of water and empty a drum or carboy to inside 3 mm of the bottom.
PE, Poly-hand pump.
  • No pressure build-up in the vessel
  • Excellent chemical resistance
  • Ergonomic handle