Scraper, SP Bel-Art
Lieferant: Bel-Art Products, a Part of SP
H368200000
BELAH368200000EA 202 EUR
BELAH368200000
Scraper, SP Bel-Art
Schaber
Polyamide, glass-reinforced. Rigid and strong one piece scraper for heavy use.
  • Flat blade with chisel edge
  • Blade may be resharpened to restore chisel edge