Mini mortar with pestle, SP Bel-Art
Lieferant: Bel-Art Products, a Part of SP
H372600100
BELAH372600100EA 1570 EUR
BELAH372600100
Mini mortar with pestle, SP Bel-Art
Mörser und Pistille Mörser
Freeze and process small tissue samples within a microcentrifuge tube. Stainless steel mortar has a welded fixture to hold a 1,5ml microcentrifuge tube. The mortar is suspended above a stainless steel cavity which holds the liquid nitrogen and is nested in a heavily insulated blue polyethylene housing.
  • Prevents tube from cracking by eliminating direct contact with liquid nitrogen
  • Includes 12,7mm long stainless steel pestle with 102×19mm polyethylene handle
  • Suction feet keep it stable on the workbench