SP Bel-Art® liquid sampler
Lieferant: Bel-Art Products, a Part of SP
H378270000
BELAH378270000EA 828 EUR
BELAH378270000
SP Bel-Art® liquid sampler
Probenehmer Probenehmer für Flüssigkeiten
Liquid sampler made of nickel-plated brass cup and tubular handle with finger grip. Cup is not replaceable and can not detach.
Suitable for immersion into a can, bottle, tank, vat ...