SP Bel-Art® three zone owder samplers
Lieferant: Bel-Art Products, a Part of SP
H378290000
BELAH378290000EA 659 EUR
BELAH378290000
SP Bel-Art® three zone owder samplers
Probenehmer Probenehmer für Feststoffe
Three zone powder samplers.
  • Each of the three cells gathers samples
  • Fine point to sample the bag contents
  • Minimum damages to the bag
Used in non-homogenous powders, pellets, and other friable materials.