Dry box gloves, neoprene, SP Bel-Art
Lieferant: Bel-Art Products, a Part of SP
H500290000 H500290010
BELAH500290000EA 948 EUR
BELAH500290000 BELAH500290010
Dry box gloves, neoprene, SP Bel-Art
Handschuhe
These neoprene gauntlets protect the user from penetration by liquid, gas or solid substances.
  • Seamless one-piece construction, made of bonded neoprene
  • Requires clamping rings to hold gloves in place