Sieve homogenisers, Cellector™
Lieferant: BELLCO GLASS
1985-85000
BELC1985-85000EA 529 EUR
BELC1985-85000
Sieve homogenisers, Cellector™
Homogenisatoren Tischhomogenisatoren
Cellector™ homogenisers, made of stainless steel, are for preparation of cell suspensions and gel extraction of proteins.
Lieferumfang: Cellector™ complete includes a pan, glass pestle, key and nine screens (one each) 10 to 150 mesh size.