Anaerobic culture tube, Hungate type
Lieferant: BELLCO GLASS
2047-16125
BELC2047-16125EA 1750 EUR
BELC2047-16125
Anaerobic culture tube, Hungate type
Röhrchen Kulturröhrchen
These tubes made of type 1 class B borosilicate glass for growing and maintaining anaerobic bacteria, culture condition.
  • Very good temperature resistance
  • Very good chemical resistance