Digital refractometers, RFM300-T
Lieferant: BELLINGHAM STANLEY
19-30 19-40
BELL19-30EA 11076 EUR
BELL19-30 BELL19-40
Digital refractometers, RFM300-T
Refraktometer Tischrefraktometer
These high precision refractometers are designed for applications in food and beverage, including fruit juice, emulsions or chemicals.
  • Flat prism surface, touch screen and Peltier control
  • Smart temperature stability check only displays results when sample is stable
  • Up to 8000 readings stored in memory can be downloaded to a PC using free downloadable software
Standards: US/EP/BP/JP. FDA 21 CFR part 11. Validation documentation IQ/OQ/PQ is available on request.
Lieferumfang: Supplied with power cable and multi-language CD manual. EU, UK and US plugsMore Product Information