Alcohol-by-volume (ABV) kit, Eclipse
Lieferant: BELLINGHAM STANLEY
44-839
BELL44-839EA 68 EUR
BELL44-839
Alcohol-by-volume (ABV) kit, Eclipse
Aräometer
This alcohol-by-volume (ABV) kit combines a refractometer and a hydrometer for the determination of alcohol content of finished beers wine and cider as well as for monitoring ferment in bottle beers and sparkling wines.
  • Quick and easy process to carry out
  • Special hydrometer scale covers most samples
  • Internet calculator via PC or Web enabled mobile device
The alcohol content of a beer, wine and cider may easily be established by combining the results of two simple test measurements, that of a refractometer (RI-Zeiss) and a hydrometer (SG). This kit is able to achieve an alcohol by volume accuracy of 0,5% ABV.
Standards: Calibration showing traceability to ICUMSA and NIST.
Bestellinformation: Each components needs to be ordered separately to for a ABV kit.
Lieferumfang: Supplied with paper lookup table for the calculation of results.
Bitte beachten Due to the achievable accuracy of this measurement system, results may not be suitable for duty or tax declaration purposes.