Calibration materials for digital density meters
Lieferant: BELLINGHAM STANLEY
90-106 90-101 90-102 90-103 90-104 90-105 90-107 90-108 90-109 90-110 90-111 90-112 90-113 90-115 90-116
BELL90-106EA 170 EUR
BELL90-106 BELL90-101 BELL90-102 BELL90-103 BELL90-104 BELL90-105 BELL90-107 BELL90-108 BELL90-109 BELL90-110 BELL90-111 BELL90-112 BELL90-113 BELL90-115 BELL90-116
Calibration materials for digital density meters
Standards Dichtestandards
Calibration materials for verifying the performance of digital density meters using the oscillating u-tube principle. Calibrated in accordance with ISO17025 and supplied in 10 ml sealed ampoules for integrity.
  • Long shelf life
  • Low uncertainty (5 d.p.)
  • Traceable to international standards (NIST)
  • UKAS certified
Density meter reference fluid calibrated in accordance with ISO17025 and supplied in a 10 ml single use, sealed glass ampoule with breakable nozzle to prevent contamination. Each item includes a UKAS certificate, safety data sheet (SDS) and comes packed in a robust aluminium tube for additional protection during shipping and for ease of storage. Density meter reference fluid has a shelf life of 3 years and can be stored at room temperature. Reference materials are supplied in a variety of densities depending on your application.
Standards: Manufactured for Bellingham + Stanley in accordance with ISO/IEC 17025:2017 by H&D Fitzgerald Ltd., a UKAS Calibration Laboratory No. 0291.
Bitte beachten Observe SDS data for opening, use and waste.