Calibration materials for digital density meters
Lieferant: BELLINGHAM STANLEY
Calibration materials for verifying the performance of digital density meters using the oscillating u-tube principle. Calibrated in accordance with ISO17025 and supplied in 10 ml sealed ampoules for integrity.
- Long shelf life
- Low uncertainty (5 d.p.)
- Traceable to international standards (NIST)
- UKAS certified
Density meter reference fluid calibrated in accordance with ISO17025 and supplied in a 10 ml single use, sealed glass ampoule with breakable nozzle to prevent contamination. Each item includes a UKAS certificate, safety data sheet (SDS) and comes packed in a robust aluminium tube for additional protection during shipping and for ease of storage. Density meter reference fluid has a shelf life of 3 years and can be stored at room temperature. Reference materials are supplied in a variety of densities depending on your application.
Standards: Manufactured for Bellingham + Stanley in accordance with ISO/IEC 17025:2017 by H&D Fitzgerald Ltd., a UKAS Calibration Laboratory No. 0291.
Bitte beachten Observe SDS data for opening, use and waste.