Bis(triphenylphosphin)palladium(II)chlorid

Lieferant: BEPHARM

Synonyme: Bis(triphenylphosphine)palladium(II) dichloride

B18251.10G
BEPHB18251.10GEA 0 EUR
BEPHB18251.10G
Bis(triphenylphosphin)palladium(II)chlorid
Bis(triphenylphosphin)palladium(II)chlorid
Formel: C₃₆H₃₀Cl₂P₂Pd
Molecular Weight: 701,91 g/mol
MDL: MFCD00009593
CAS-Nummer: 13965-03-2
EINECS: 237-744-2

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR