2-Brom-9-fluorenon

Lieferant: BEPHARM

Synonyme: 2-Bromo-9H-fluoren-9-one

B20131-2000G
BEPHB20131-2000GEA 0 EUR
BEPHB20131-2000G
2-Brom-9-fluorenon
2-Brom-9-fluorenon
Formel: C₁₃H₇BrO
Molecular Weight: 259,1 g/mol
MDL: MFCD00010789
CAS-Nummer: 3096-56-8

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR